Learn More
Cayman Chemical CalceIn UltraBlueAM 50ug
SDP
Catalog No. 502984582
Encompass_Preferred
A fluorogenic dye used to assess cell viability; ex/em maxima = 360/445 nm, respectively
Catalog No. 50-298-4582 Supplier Cayman Chemical Supplier No. 2063950 UG
A fluorogenic dye used to assess cell viability; ex/em maxima = 360/445 nm, respectively