Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcein Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium. | 154071-48-4 | 25G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C087525G
Encompass
Calcein Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium. | Mol Wt: 622.53 | 154071-48-4 | 25G
Catalog No. 50-186-4003