Learn More
Medchemexpress LLC Calceolarioside B | 105471-98-5 | 478.45 | 5 MG
SDP
Catalog No. 5000388501
Encompass_Preferred
Calceolarioside B is a phenylethanoid glycoside that can be isolated from the leaves of Akebia quinata. It inhibits RLAR activity with an IC50 value of 23.99 μM, inhibits the entry of the Omicron BA.2 pseudovirus into host cells, and reduces IL-6 levels. Calceolarioside B also possesses immunomodulatory and anticancer activities against human hormone-independent prostate cancer.
  • Inhibits RLAR activity.
  • Inhibits entry of Omicron BA.2 pseudovirus into host cells.
  • Reduces IL-6 levels.
  • Possesses immunomodulatory activity.
  • Exhibits anticancer activity against human hormone-independent prostate cancer.
Catalog No. 50-003-88501 Supplier Medchemexpress LLC Supplier No. HYN05395MG
Calceolarioside B is a phenylethanoid glycoside that can be isolated from the leaves of Akebia quinata. It inhibits RLAR activity with an IC50 value of 23.99 μM, inhibits the entry of the Omicron BA.2 pseudovirus into host cells, and reduces IL-6 levels. Calceolarioside B also possesses immunomodulatory and anticancer activities against human hormone-independent prostate cancer.
  • Inhibits RLAR activity.
  • Inhibits entry of Omicron BA.2 pseudovirus into host cells.
  • Reduces IL-6 levels.
  • Possesses immunomodulatory activity.
  • Exhibits anticancer activity against human hormone-independent prostate cancer.