Learn More
Apexbio Technology LLC Calcifediol, 25mg. Cas: 19356-17-3 MFCD: MFCD00867077. Major circulating form of vitamin D3
SDP
Supplier:  Apexbio Technology LLC B214025
Encompass_Preferred
Vitamin D receptor (VDR) activation is associated with cardiovascular and survival benefits in chronic kidney disease patients. Calcifediol, also known as calcidiol, is a prehormone that is produced in the liver by hydroxylation of vitamin D3. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5315
 Restricted Item