Learn More
Medchemexpress LLC HY-32351 5mg Medchemexpress, Calcifediol CAS:19356-17-3 Purity:>98%
SDP
Catalog No. 502082230
Encompass_Preferred
Medchemexpress, HY-32351 5mg Calcifediol CAS:19356-17-3 Calcifediol (25-hydroxy Vitamin D3), a major circulating metabolite of vitamin D3, is a potent VDR inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2230 Supplier Medchemexpress LLC Supplier No. HY323515MG
Medchemexpress, HY-32351 5mg Calcifediol CAS:19356-17-3 Calcifediol (25-hydroxy Vitamin D3), a major circulating metabolite of vitamin D3, is a potent VDR inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.