Learn More
Medchemexpress LLC Calcifediol-d3 | 140710-94-7 | 99.0% | 403.66 | 1 MG
SDP
Catalog No. 5000401479
Encompass_Preferred
Calcifediol-d3 is a deuterium labeled Calcifediol. Calcifediol is an effective VDR ligand and VD supplement, serving as a prohormone of the vitamin D endocrine system (VDES). It is hydroxylated in the liver to produce the active form, calcitriol, and can rapidly increase serum VD levels.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-01479 Supplier Medchemexpress LLC Supplier No. HY32351S1MG
Calcifediol-d3 is a deuterium labeled Calcifediol. Calcifediol is an effective VDR ligand and VD supplement, serving as a prohormone of the vitamin D endocrine system (VDES). It is hydroxylated in the liver to produce the active form, calcitriol, and can rapidly increase serum VD levels.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.