Learn More
Medchemexpress LLC Calcifediol-d6 (25-hydroxy vitamin D3-d6) | 78782-98-6 | MFCD00870231 | 99.1% | 406.67 | C27H38D6O2 | 5 MG
SDP
Catalog No. 5000262626
Encompass_Preferred
Calcifediol-d6 is the deuterium-labeled analogue of calcifediol (25-hydroxy vitamin D3), a major circulating metabolite and potent vitamin D receptor (VDR) ligand. The isotopically labeled compound is provided as a high-purity analytical standard for quantitative mass spectrometry, tracer studies, and related research applications.
  • Deuterated internal standard for LC-MS/MS quantification.
  • High purity suitable for analytical use (>99%).
  • Molecular formula C27H38D6O2; molecular weight 406.67.
  • Supplied as a solid for solution preparation and storage.
  • Suitable for pharmacokinetic, metabolic, and receptor-binding studies.
Catalog No. 50-002-62626 Supplier Medchemexpress LLC Supplier No. HY133325MG
Calcifediol-d6 is the deuterium-labeled analogue of calcifediol (25-hydroxy vitamin D3), a major circulating metabolite and potent vitamin D receptor (VDR) ligand. The isotopically labeled compound is provided as a high-purity analytical standard for quantitative mass spectrometry, tracer studies, and related research applications.
  • Deuterated internal standard for LC-MS/MS quantification.
  • High purity suitable for analytical use (>99%).
  • Molecular formula C27H38D6O2; molecular weight 406.67.
  • Supplied as a solid for solution preparation and storage.
  • Suitable for pharmacokinetic, metabolic, and receptor-binding studies.