Learn More
Medchemexpress LLC Calcifediol-d6 | 78782-98-6 | MFCD15071183 | 99.1% | C27H38D6O2 | 10MG
SDP
Catalog No. 5000258425
Encompass_Preferred
Calcifediol-d6 is the deuterium-labeled form of calcifediol (25-hydroxyvitamin D3), supplied as an isotopically labeled internal standard for analytical and research applications. It is intended for quantitative mass spectrometry, metabolic profiling, and receptor binding studies, where a close mass match to the native analyte improves accuracy and precision.
  • Deuterium-labeled internal standard for LC-MS/MS quantification.
  • High isotopic purity for accurate mass resolution.
  • Reported purity 99.1%.
  • Molecular formula C27H38D6O2 and molecular weight ~406.7 g/mol.
  • Available in multiple pack sizes suitable for analytical workflows.
Catalog No. 50-002-58425 Supplier Medchemexpress LLC Supplier No. HY1333210MG
Calcifediol-d6 is the deuterium-labeled form of calcifediol (25-hydroxyvitamin D3), supplied as an isotopically labeled internal standard for analytical and research applications. It is intended for quantitative mass spectrometry, metabolic profiling, and receptor binding studies, where a close mass match to the native analyte improves accuracy and precision.
  • Deuterium-labeled internal standard for LC-MS/MS quantification.
  • High isotopic purity for accurate mass resolution.
  • Reported purity 99.1%.
  • Molecular formula C27H38D6O2 and molecular weight ~406.7 g/mol.
  • Available in multiple pack sizes suitable for analytical workflows.