Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcifediol, Tocris Bioscience™

Catalog No. 40361
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-361 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-361 Supplier R&D Systems Supplier No. 4036/1
This item is not returnable. View return policy

Prohormone of calcitriol (Cat. No. 2551). Major circulating form of vitamin D

Prohormone of calcitriol (Cat. No. 2551). Major circulating form of vitamin D. Suppresses PTH synthesis and secretion in bovine parathyroid cells (ED50 = 2 nM).

Chemical Identifiers

CAS 19356-17-3
Molecular Formula C27H44O2
Molecular Weight (g/mol) 400.65
MDL Number MFCD00867077,MFCD11656130,MFCD11656128
InChI Key JWUBBDSIWDLEOM-DTOXIADCSA-N
Synonym (εR,1R,3aS,4E,7aR)-4-[(2Z)-2-[(5S)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]octahydro-α,α,ε,7a-tetramethyl-1H-indene-1-pentanol
PubChem CID 126968450
IUPAC Name (1S,3Z)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)CCC1=C

Specifications

Quantity 1 mg
Formula Weight 400.64
Percent Purity >98%
Chemical Name or Material Calcifediol
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.