Learn More
Medchemexpress LLC Calcineurin A Antibody | 10 UL
SDP
Catalog No. 5000443611
Encompass_Preferred
Calcineurin A Antibody (YA2508) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Calcineurin A. It is suitable for research use only and plays a crucial role in transducing intracellular Ca(2+)-mediated signals. Its molecular weight is observed and predicted at 59 kDa.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated and unmodified
  • Targeted to Calcineurin A
  • Suitable for Western Blot, Immunocytochemistry/Immunofluorescence, and Flow Cytometry applications
  • Recognizes human, mouse, and rat species
  • Purified by Affinity Chromatography
Catalog No. 50-004-43611 Supplier Medchemexpress LLC Supplier No. HYP8276310UL
Calcineurin A Antibody (YA2508) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Calcineurin A. It is suitable for research use only and plays a crucial role in transducing intracellular Ca(2+)-mediated signals. Its molecular weight is observed and predicted at 59 kDa.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated and unmodified
  • Targeted to Calcineurin A
  • Suitable for Western Blot, Immunocytochemistry/Immunofluorescence, and Flow Cytometry applications
  • Recognizes human, mouse, and rat species
  • Purified by Affinity Chromatography