Learn More
Enzo Life Sciences Calcineurin A polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA610F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to human calcineurin A. UniProt ID: P16298. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Drosophila, Dog, Chicken, Bovine. Applications: IP, WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7451