Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcineurin a antibody (YA2508) | 100 UL
SDP

Catalog No. 5000444511
Encompass_Preferred

Calcineurin A Antibody (YA2508) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Calcineurin A. It is suitable for Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC) applications in human, mouse, and rat samples.

  • Derived from rabbit.
  • Monoclonal antibody.
  • Isotype is IgG.
  • Applications include western blot (WB), immunocytochemistry/immunofluorescence (ICC/IF), and flow cytometry (FC).
  • Reacts with human, mouse, and rat samples.

Catalog No. 50-004-44511 Supplier Medchemexpress LLC Supplier No. HYP82763100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcineurin A Antibody (YA2508) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Calcineurin A. It is suitable for Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), and Flow Cytometry (FC) applications in human, mouse, and rat samples.

  • Derived from rabbit.
  • Monoclonal antibody.
  • Isotype is IgG.
  • Applications include western blot (WB), immunocytochemistry/immunofluorescence (ICC/IF), and flow cytometry (FC).
  • Reacts with human, mouse, and rat samples.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.