Learn More
Enzo Life Sciences Calcineurin Phosphatase assay kit (1x96wells)
SDP
Catalog No. 502010301
Encompass_Preferred
This colorimetric kit is designed for inhibitor screening or kinetic assay of recombinant Calcineurin. Activity is determined using the RII phosphopeptide substrate, the most efficient and selective peptide known for calcineurin, and detection of free phosphate released is based on the classic malachite green assay. Applications: Colorimetric detection, HTS Activity assay.
Catalog No. 50-201-0301 Supplier Enzo Life Sciences Supplier No. BMLAK8040001
Add to Cart
Add to Cart
This colorimetric kit is designed for inhibitor screening or kinetic assay of recombinant Calcineurin. Activity is determined using the RII phosphopeptide substrate, the most efficient and selective peptide known for calcineurin, and detection of free phosphate released is based on the classic malachite green assay. Applications: Colorimetric detection, HTS Activity assay.