Learn More
Medchemexpress LLC Calcineurin autoinhibitory peptide | 148067-21-4 | 99.99% | 2930.32 | 1 MG
SDP
Catalog No. 5000423913
Encompass_Preferred
Calcineurin autoinhibitory peptide is a selective inhibitor of Ca2+/calmodulin-dependent protein phosphatase (calcineurin), with an IC50 of approximately 10 μM. It has been shown to protect neurons from excitatory neuronal death, making it suitable for various research applications in neurobiology.
  • Selective inhibitor of Ca2+/calmodulin-dependent protein phosphatase (calcineurin)
  • IC50 of approximately 10 μM
  • Protects neurons from excitatory neuronal death
Catalog No. 50-004-23913 Supplier Medchemexpress LLC Supplier No. HYP12471MG
Calcineurin autoinhibitory peptide is a selective inhibitor of Ca2+/calmodulin-dependent protein phosphatase (calcineurin), with an IC50 of approximately 10 μM. It has been shown to protect neurons from excitatory neuronal death, making it suitable for various research applications in neurobiology.
  • Selective inhibitor of Ca2+/calmodulin-dependent protein phosphatase (calcineurin)
  • IC50 of approximately 10 μM
  • Protects neurons from excitatory neuronal death