Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcineurin autoinhibitory peptide | 148067-21-4 | 99.99% | 2930.32 | 5 MG
SDP

Catalog No. 5000424325
Encompass_Preferred

Calcineurin autoinhibitory peptide is a selective inhibitor of Ca2+/calmodulin-dependent protein phosphatase (calcineurin), with an IC50 of approximately 10 μM. It can protect neurons from excitatory neuronal death.

  • Selective inhibitor of calcineurin
  • Protects neurons from excitatory neuronal death
  • Target: Phosphatase
  • Pathway: Metabolic Enzyme/Protease
  • Optimal storage for powder: -80°C for 2 years or -20°C for 1 year (sealed, away from moisture)
  • Optimal storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • Soluble in H2O at 100 mg/mL (34.13 mM)
  • Soluble in PBS at 100 mg/mL (34.13 mM)

Catalog No. 50-004-24325 Supplier Medchemexpress LLC Supplier No. HYP12475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Calcineurin autoinhibitory peptide is a selective inhibitor of Ca2+/calmodulin-dependent protein phosphatase (calcineurin), with an IC50 of approximately 10 μM. It can protect neurons from excitatory neuronal death.

  • Selective inhibitor of calcineurin
  • Protects neurons from excitatory neuronal death
  • Target: Phosphatase
  • Pathway: Metabolic Enzyme/Protease
  • Optimal storage for powder: -80°C for 2 years or -20°C for 1 year (sealed, away from moisture)
  • Optimal storage in solvent: -80°C for 6 months or -20°C for 1 month (sealed, away from moisture)
  • Soluble in H2O at 100 mg/mL (34.13 mM)
  • Soluble in PBS at 100 mg/mL (34.13 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.