Learn More
Medchemexpress LLC HY-P0228A 1mg Medchemexpress, Calcineurin substrate (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0228A1MG
Encompass_Preferred
Medchemexpress, HY-P0228A 1mg Calcineurin substrate (TFA) CAS: Calcineurin substrate (TFA) is a peptide from the regulatory RII subunit of cAMP-dependent protein kinase. Calcineurin substrate (TFA) can be used in the calcineurin activity assay [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4062