Learn More
Cayman Chemical CalcInurIn SubstrATtrIflu 10mg
SDP
Catalog No. 502951024
Encompass_Preferred
A synthetic peptide substrate; can be used as a substrate to quantify calcineurin activity
Catalog No. 50-295-1024 Supplier Cayman Chemical Supplier No. 3467010 MG
A synthetic peptide substrate; can be used as a substrate to quantify calcineurin activity