Learn More
Medchemexpress LLC HY-10001 10mg Medchemexpress, Calcipotriol CAS:112965-21-6 Purity:>98%
SDP
Catalog No. 502028599
Encompass_Preferred
Medchemexpress, HY-10001 10mg Calcipotriol CAS:112965-21-6 Calcipotriol is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8599 Supplier Medchemexpress LLC Supplier No. HY1000110MG
Add to Cart
Add to Cart
Medchemexpress, HY-10001 10mg Calcipotriol CAS:112965-21-6 Calcipotriol is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.