Learn More
Medchemexpress LLC Calcipotriol | 112965-21-6 | MFCD10567086 | 99.4% | 412.60 g·mol-1 | C27H40O3 | 10 MG
SDP
Catalog No. 5000215342
Encompass_Preferred
Calcipotriol (Standard) is an analytical reference standard of the vitamin D3 analogue calcipotriol (calcipotriene), intended for research and analytical applications. It is provided as a high-purity material suitable for chromatographic reference, method development, and quantitative assays. Molecular formula C27H40O3, molecular weight 412.60 g·mol-1, CAS 112965-21-6.
  • High purity suitable for analytical reference and quantitative assays.
  • Compatible with HPLC and other chromatographic methods.
  • Available in small pack sizes for standards, including 10 MG.
  • Molecular formula and molecular weight provided for accurate calculations.
Catalog No. 50-002-15342 Supplier Medchemexpress LLC Supplier No. HY10001R10MG
Calcipotriol (Standard) is an analytical reference standard of the vitamin D3 analogue calcipotriol (calcipotriene), intended for research and analytical applications. It is provided as a high-purity material suitable for chromatographic reference, method development, and quantitative assays. Molecular formula C27H40O3, molecular weight 412.60 g·mol-1, CAS 112965-21-6.
  • High purity suitable for analytical reference and quantitative assays.
  • Compatible with HPLC and other chromatographic methods.
  • Available in small pack sizes for standards, including 10 MG.
  • Molecular formula and molecular weight provided for accurate calculations.