Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calcipotriol Impurity C | 113082-99-8 | 99.1% | C27H40O3 | 5 MG
SDP

Catalog No. 5000434656
Encompass_Preferred

Calcipotriol Impurity C is an impurity of Calcipotriol, which functions as a ligand for VDR-like receptors. This product is intended for research use only.

  • Functions as a ligand for VDR-like receptors
  • Target is VDR (Vitamin D Receptor)
  • Soluble in DMSO
  • Soluble in 10% DMSO >> 90% (20% SBE-β-CD in saline)
  • Soluble in 10% DMSO >> 90% corn oil
  • Store at 4°C, protected from light, under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen

Catalog No. 50-004-34656 Supplier Medchemexpress LLC Supplier No. HY750355MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Calcipotriol Impurity C is an impurity of Calcipotriol, which functions as a ligand for VDR-like receptors. This product is intended for research use only.

  • Functions as a ligand for VDR-like receptors
  • Target is VDR (Vitamin D Receptor)
  • Soluble in DMSO
  • Soluble in 10% DMSO >> 90% (20% SBE-β-CD in saline)
  • Soluble in 10% DMSO >> 90% corn oil
  • Store at 4°C, protected from light, under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light, under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.