Learn More
Medchemexpress LLC Calcipotriol impurity C | 113082-99-8 | MFCD22573293 | 99.2% | 412.6 g/mol | C27H40O3 | 10 MG
SDP
Catalog No. 5000299179
Encompass_Preferred
Calcipotriol impurity C is an analytical reference standard and a known impurity of calcipotriol, provided for research and analytical use in vitamin D-related studies. It is supplied as a solid standard for laboratory method development and impurity identification; it is not validated for medical applications.
  • Analytical reference for impurity identification and method development.
  • Intended for research use only; not validated for medical applications.
  • Molecular formula C27H40O3; molecular weight 412.6 g/mol.
  • Available as a 10 mg solid; additional sizes available for analytical needs.
  • Store protected from light and under nitrogen; in solvent: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-99179 Supplier Medchemexpress LLC Supplier No. HY7503510MG
Calcipotriol impurity C is an analytical reference standard and a known impurity of calcipotriol, provided for research and analytical use in vitamin D-related studies. It is supplied as a solid standard for laboratory method development and impurity identification; it is not validated for medical applications.
  • Analytical reference for impurity identification and method development.
  • Intended for research use only; not validated for medical applications.
  • Molecular formula C27H40O3; molecular weight 412.6 g/mol.
  • Available as a 10 mg solid; additional sizes available for analytical needs.
  • Store protected from light and under nitrogen; in solvent: -80°C up to 6 months, -20°C up to 1 month.