Learn More
Medchemexpress LLC HY-10001A 5mg Medchemexpress, Calcipotriol (monohydrate) CAS:147657-22-5 Purity:>98%
SDP
Catalog No. 501931693
Encompass_Preferred
Medchemexpress, HY-10001A 5mg Calcipotriol (monohydrate) CAS:147657-22-5 Calcipotriol monohydrate is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1693 Supplier Medchemexpress LLC Supplier No. HY10001A5MG
Medchemexpress, HY-10001A 5mg Calcipotriol (monohydrate) CAS:147657-22-5 Calcipotriol monohydrate is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.