Learn More
Bachem Calcitonin (8-32) (salmon I) Trifluoroacetate
SDP
Supplier:  Bachem 4037182.1000
Encompass_Preferred
Calcitonin (8-32) (salmon I) Trifluoroacetate, H-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Gly-Thr-Pro-NH2, 155069-90-2, C119H198N36O37, Mr 2725.10, 1mg. Calcitonin (8-32) (salmon I) has been been proposed as an amylin receptor antagonist which is highly selective for amylin. sCT 8-32 reverses the inhibitory effect of amylin on glucose-induced insulin release and potentiates the insulin response to glucose in the rat perfused pancreas.
Catalog No. 50-194-6907