Learn More
Bachem Calcitonin C-Terminal Flanking Peptide (human) Acetate
SDP
Supplier:  Bachem 4102089.1000
Encompass_Preferred
Calcitonin C-Terminal Flanking Peptide (human) Acetate, H-Asp-Met-Ser-Ser-Asp-Leu-Glu-Arg-Asp-His-Arg-Pro-His-Val-Ser-Met-Pro-Gln-Asn-Ala-Asn-OH, 85916-47-8, C97H154N34O36S2, Mr 2436.63, 1mg. C-Procalcitonin (human), Katacalcin, PDN-21.
Catalog No. 50-194-7443