Learn More
Medchemexpress LLC Calcitonin/CALCA Mo 100ug | 100ug
SDP
Catalog No. 5000187229
Encompass_Preferred
The Calcitonin/CALCA protein induces vasodilation in different blood vessels including the coronary cerebral and systemic vasculature It is highly concentrated in the central nervous system suggesting a role as a neurotransmitter or neuromodulator Moreover it increases platelet cAMP levels possibly affecting platelet function Calcitonin/CALCA Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Calcitonin/CALCA protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-87229 Supplier Medchemexpress LLC Supplier No. HYP77587100UG
The Calcitonin/CALCA protein induces vasodilation in different blood vessels including the coronary cerebral and systemic vasculature It is highly concentrated in the central nervous system suggesting a role as a neurotransmitter or neuromodulator Moreover it increases platelet cAMP levels possibly affecting platelet function Calcitonin/CALCA Protein Mouse (HEK293 Fc) is the recombinant mouse-derived Calcitonin/CALCA protein expressed by HEK293 with C-hFc labeled tag