Learn More
Medchemexpress LLC HY-P1913A 5mg Medchemexpress, Calcitonin Gene Related Peptide (CGRP) II, rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1913A5MG
Encompass_Preferred
Medchemexpress, HY-P1913A 5mg Calcitonin Gene Related Peptide (CGRP) II, rat (TFA) CAS: Calcitonin Gene Related Peptide (CGRP) II, rat (TFA) is a neuropeptide with 37 amino acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3077