Learn More
CPC Scientific H-Ala-Cys-Asp-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2
(Disulfide bridge) 0.5MG
SDP
Supplier:  CPC Scientific OSTP018A
Encompass
Calcitonin gene-related peptide (CGRP) is part of the calcitonin family of peptides, which in humans exists in two forms, alpha-CGRP and beta-CGRP. alpha-CGRP is a 37-amino acid peptide formed from the alternative genetic splicing of the calcitonin/CGRP geneSEQUENCE: H-Ala-Cys-Asp-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2(Disulfide bridge)ONE-LETTER SEQUENCE: ACDTATCVTHRLAGLLSRSGGVVKNNFVPTNVGSKAF-NH2 (C2&C7 bridge)MOLECULAR FORMULA: C163H267N51O49S2MOLECULAR WEIGHT: 3789.36STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [90954-53-3]SYNONYMS: CGRP-I (human), Calcitonin Gene-Related Peptide-1RESEARCH AREA: CardiovascularREFERENCES:P.Le Greves et al., Eur. J. Pharmacol., 115, 309 (1985)H.R.Morris et al., Nature, 308, 746 (1984)
SKU(s): OSTP-018A, OSTP-018B
Catalog No. 50-211-0031