Learn More
CPC Scientific H-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asn-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OSTP020B
Encompass
Selective antagonist for CGRP receptors (but not for calcitonin receptors) that cross-reacts with each other at the level of their distinct receptors, thereby is a valuable tool for receptor characterization studies. ONE-LETTER SEQUENCE: VTHRLAGLLSRSGGVVKNNFVPTNVGSKAF-NH2MOLECULAR FORMULA: C139H230N44O38MOLECULAR WEIGHT:3125.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [119911-68-1], RESEARCH AREA: OsteoporosisREFERENCES: T. Chiba et al., Amer. J. Physiol., 256, E331 (1989)
Catalog No. 50-211-0036