Learn More
CPC Scientific H-Ser-Cys-Asn-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asp-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Glu-Ala-Phe-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific OSTP023A
Encompass
CGRP induces elevation of cAMP and cGMP in rat aorta.ONE-LETTER SEQUENCE: SCNTATCVTHRLAGLLSRSGGVVKDNFVPTNVGSEAF-NH2 (Cys2 and 7 bridge)MOLECULAR FORMULA: C162H262N50O52S2MOLECULAR WEIGHT:3806.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [96827-03-1], SYNONYMS: CGRP (rat), Calcitonin Gene Related Peptide-1, alpha-CGRP (rat)RESEARCH AREA: CardiovascularREFERENCES: S.G. Amara, V. Jonas, M.G. Rosenfeld, E.S. Ong and R.M. Evans, Nature, 298, 240 (1982) (Original); M.G. Rosenfeld, J.J. Mermond, S.G. Amara, L.W. Swanson, P.E. Sawchenko, J. Rivier, W.W. Vale and R.M. Evans, Nature, 304, 129 (1983) (Processing and Distribution in Neural Tissue)
Catalog No. 50-211-0041