Learn More
Genscript Corporation Calcitonin Gene Related Peptide (CGRP) (8-37), rat
Supplier:  Genscript Corporation RP110900.5
Encompass
Calcitonin Gene Related Peptide (CGRP)(8-37) inhibits the evoked discharge frequency of wide dynamic range neurons in dorsal horn of the spinal cord in rats. The present study was performed to explore the effect of CGRP(8-37) on the electrical stimulation-evoked discharge frequency of wide dynamic range (WDR) neurons in the dorsal horn of the spinal cord in rats. The results indicate that CGRP receptors play an important role in the transmission of presumed nociceptive information in the dorsal horn of the spinal cord.
Catalog No. 50-275-009