Learn More
Eurofins Discoverx Calcitonin, human, 1 mg. (92-1054)
Supplier:  Eurofins Discoverx 921054
Encompass
Calcitonin is a polypeptide hormone produced in the thyroid by parafollicular cells (C-Cells). It reduces blood calcium levels in opposition to the effects of parathyroid hormone. It participates in both calcium and phosphorus metabolism, stimulates osteoblast activity, and inhibits intestinal calcium absorption, osteoclast activity, and renal tubular cell calcium absorption. Calcitonin levels in the blood are dictated by blood calcium levels. Acts as an agonist for CALCR. Provided as 1 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9595