Learn More
Carbosynth LLC. Calcitonin (Human) (0.5 mg vial), 1ea, Disulfide bonds: (Disulfide bond between Cys1-Cys7), MW: 3417.8, Molecular formula: C151H226N40O45S3, CAS# 21215-62-3
SDP
Supplier:  Carbosynth LLC. PCL4051V0.5MG
Encompass_Preferred
Synonyms: Cys-Gly-Asn-Leu-Ser-Thr-Cys-Met-Leu-Gly-Thr-Tyr-Thr-Gln-Asp-Phe-Asn-Lys-Phe-His-Thr-Phe-Pro-Gln-Thr-Ala-Ile-Gly-Val-Gly-Ala-Pro-NH2, Storage Temperature: -20°C, References: R. Neher, B. Riniker, W. Rittel, and H. Zuber, Helv. Chim. Acta, 51, 1900 (1968). (Original), Y. Nakagawa, T. Morikawa, and S. Sakakibara, Peptide Chemistry 1977, 189 (1978). (Chem. Synthesis)
Catalog No. 50-168-6374