Learn More
Medchemexpress LLC Calcitonin (salmon) | 47931-85-1 | 99.96% | 3431.85 | 10 MG
SDP
Catalog No. 5000425745
Encompass_Preferred
Calcitonin (salmon) is a calcium regulating hormone and a dual-action amylin and calcitonin receptor agonist. It stimulates bone formation and inhibits bone resorption. Oral treatment with Calcitonin (salmon) dose-dependently attenuates fasting and non-fasted hyperglycemia, reduces diabetic hyperglycemia and HbA1c levels, and improves pancreatic beta-cell function and area.
  • Stimulates bone formation
  • Inhibits bone resorption
  • Attenuates fasting and non-fasted hyperglycemia
  • Reduces diabetic hyperglycemia
  • Reduces HbA1c levels
  • Improves pancreatic beta-cell function
  • Improves pancreatic beta-cell area
  • Acts as a dual-action amylin and calcitonin receptor agonist
Catalog No. 50-004-25745 Supplier Medchemexpress LLC Supplier No. HYP009010MG
Calcitonin (salmon) is a calcium regulating hormone and a dual-action amylin and calcitonin receptor agonist. It stimulates bone formation and inhibits bone resorption. Oral treatment with Calcitonin (salmon) dose-dependently attenuates fasting and non-fasted hyperglycemia, reduces diabetic hyperglycemia and HbA1c levels, and improves pancreatic beta-cell function and area.
  • Stimulates bone formation
  • Inhibits bone resorption
  • Attenuates fasting and non-fasted hyperglycemia
  • Reduces diabetic hyperglycemia
  • Reduces HbA1c levels
  • Improves pancreatic beta-cell function
  • Improves pancreatic beta-cell area
  • Acts as a dual-action amylin and calcitonin receptor agonist