Learn More
Thermo Scientific Chemicals Calcitonin, Salmon
Description
It shows hypocalcaemic and hypophosphatemic action, with amino-acid structure differing amongst mammalian species. Salmon calcitonin (salcatonin is the synthetic analogue) shows a markedly different sequence to that of the higher vertebrates as well as more potent activity. It decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Species | Salmon |
| Purification Method | HPLC |
| Format | Powder |
| For Use With (Application) | A peptide hormone |
| Molecular Weight (g/mol) | 3417.90 |
| Product Type | Calcitonin, salmon |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.