Learn More
StatLab Calcitonin Thyroid Cancer IHC Control Slides
SDP
Catalog No. 21033329
Change view
Click to view available options
Quantity:
15 Pk.
Catalog No. Quantity
21-033-329 15 Pk.
1 options
Catalog No. 21-033-329 Supplier StatLab Supplier No. CSCALCIT/15
Add to Cart
Add to Cart
Use calcitonin thyroid cancer control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 15 Unstained Slides per pack
Specifications
Host Species Human
Quantity 15 Pk.
Product Type Control Slide