Learn More
Cayman Chemical CalcItonInel 10mg
SDP
Catalog No. 502991095
Encompass_Preferred
A peptide hormone that lowers blood calcium levels and inhibits bone resorption; binds to CTR2 with higher affinity than human, porcine, or rat calcitonin (Kis = 0.01, 2.95, 0.44, and 5.89 nM, respectively, in radioligand binding assays); inhibits gastric emptying more potently than human calcitonin in conscious dogs at 10 UG/kg; increases the paw withdrawal threshold in a mouse model of carrageenan-induced inflammation at 0.2 UG/paw
Catalog No. 50-299-1095 Supplier Cayman Chemical Supplier No. 3148710 MG
A peptide hormone that lowers blood calcium levels and inhibits bone resorption; binds to CTR2 with higher affinity than human, porcine, or rat calcitonin (Kis = 0.01, 2.95, 0.44, and 5.89 nM, respectively, in radioligand binding assays); inhibits gastric emptying more potently than human calcitonin in conscious dogs at 10 UG/kg; increases the paw withdrawal threshold in a mouse model of carrageenan-induced inflammation at 0.2 UG/paw