Learn More
Medchemexpress LLC Calcitriol (1,25-dihydroxyvitamin D3) | 32222-06-3 | MFCD00867079 | 99.9% | 416.64 g/mol | C27H44O3 | 1 MG
SDP
Catalog No. 5000215670
Encompass_Preferred
Calcitriol (1,25-dihydroxyvitamin D3) analytical standard supplied as a high-purity solid for use as a reference material in research and analytical applications. It is the most active metabolite of vitamin D and acts as a vitamin D receptor agonist, suitable for method development, calibration, and quantitative assays using chromatographic and spectrometric techniques.
  • High-purity analytical standard suitable for reference use.
  • Intended for HPLC, GC, and mass spectrometry methods.
  • Suitable for qualitative and quantitative method development and calibration.
  • Supplied in small pack sizes appropriate for analytical standards.
  • Store under recommended conditions specified in the certificate of analysis.
Catalog No. 50-002-15670 Supplier Medchemexpress LLC Supplier No. HY10002R1MG
Calcitriol (1,25-dihydroxyvitamin D3) analytical standard supplied as a high-purity solid for use as a reference material in research and analytical applications. It is the most active metabolite of vitamin D and acts as a vitamin D receptor agonist, suitable for method development, calibration, and quantitative assays using chromatographic and spectrometric techniques.
  • High-purity analytical standard suitable for reference use.
  • Intended for HPLC, GC, and mass spectrometry methods.
  • Suitable for qualitative and quantitative method development and calibration.
  • Supplied in small pack sizes appropriate for analytical standards.
  • Store under recommended conditions specified in the certificate of analysis.