Learn More
Medchemexpress LLC HY-10002 10mg , Calcitriol CAS:32222-06-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000210MG
Encompass_Preferred
HY-10002 10mg Calcitriol CAS: 32222-06-3 Calcitriol is the most active metabolite of vitamin D and also a vitamin D receptor (VDR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0344