Learn More
Medchemexpress LLC HY-10002 2mg Medchemexpress, Calcitriol CAS:32222-06-3 Purity:>98%
SDP
Catalog No. 502028604
Encompass_Preferred
Medchemexpress, HY-10002 2mg Calcitriol CAS:32222-06-3 Calcitriol is the most active metabolite of vitamin D and also a vitamin D receptor (VDR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8604 Supplier Medchemexpress LLC Supplier No. HY100022MG
Medchemexpress, HY-10002 2mg Calcitriol CAS:32222-06-3 Calcitriol is the most active metabolite of vitamin D and also a vitamin D receptor (VDR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.