Learn More
Medchemexpress LLC Calcitriol-d6 | 78782-99-7 | 99.3% | C27H38D6O3 | 5 MG
SDP
Catalog No. 5000434484
Encompass_Preferred
Calcitriol-d6 is the deuterium labeled form of Calcitriol, which is the active metabolite of vitamin D3 and an agonist of the vitamin D receptor (VDR). It is the hormonally active form of vitamin D. This compound can serve as a valuable tool in quantitative analysis.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Internal standard for quantitative analysis of Calcitriol in plasma.
Catalog No. 50-004-34484 Supplier Medchemexpress LLC Supplier No. HY768145MG
Calcitriol-d6 is the deuterium labeled form of Calcitriol, which is the active metabolite of vitamin D3 and an agonist of the vitamin D receptor (VDR). It is the hormonally active form of vitamin D. This compound can serve as a valuable tool in quantitative analysis.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Internal standard for quantitative analysis of Calcitriol in plasma.