Learn More
Medchemexpress LLC Calcitriol-d6 | 78782-99-7 | MFCD15071365 | 99.3% | 422.67 | C27H38D6O3 | 10 MG
SDP
Catalog No. 5000298157
Encompass_Preferred
Calcitriol-d6 is the deuterium-labeled form of calcitriol (1,25-dihydroxyvitamin D3) used as an isotopically labeled internal standard in research and quantitative analysis.
  • Deuterium-labeled (d6) analog of calcitriol.
  • Intended for use as an internal standard for LC-MS or GC-MS quantification.
  • High chemical purity suitable for analytical applications.
  • Molecular formula C27H38D6O3; molecular weight 422.67.
Catalog No. 50-002-98157 Supplier Medchemexpress LLC Supplier No. HY7681410MG
Calcitriol-d6 is the deuterium-labeled form of calcitriol (1,25-dihydroxyvitamin D3) used as an isotopically labeled internal standard in research and quantitative analysis.
  • Deuterium-labeled (d6) analog of calcitriol.
  • Intended for use as an internal standard for LC-MS or GC-MS quantification.
  • High chemical purity suitable for analytical applications.
  • Molecular formula C27H38D6O3; molecular weight 422.67.