Learn More
Biomatik Corporation Calcitroic Acid (CA) CLIA Kit
Supplier:  Biomatik Corporation EKU08664
Encompass_Preferred
Calcitroic Acid (CA) CLIA Kit for detecting CA in biological agents. Species Reactivity: Universal. Detection Range: 195.3-50,000pg/ml. Sensitivity: 75.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8640