Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Calciu mL-Lactate Hydrate

Catalog No. L2000250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L2000250G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L2000250G Supplier Sigma Aldrich Supplier No. L2000250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Calciu mL-lactate hydrate has been used as a sole carbon and energy source, to study the sulfate reduction ability of Desulfovibrio desulfuricans under different conditions.

Specifications

CAS 68303-40-2
Molecular Formula C4H5NaO5
Linear Formula C4H5O5Na
MDL Number MFCD00144648
Quantity 25 g
Synonym (S)-(+)-2-Hydroxypropanoic acid hemicalcium salt; L-Lactic acid hemicalcium salt; Sarcolactic acid hemicalcium salt
Molecular Weight (g/mol) 156.07 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.