Learn More
Hach Company Calcium Hardness Standard Solution, 10,000 mg/L as CaCO₃ (NIST), pk/16 10-mL Ampules
Catalog No. 501994716 Shop All Hach Company Products
Encompass
For accuracy checks in calcium hardness determinations.
Catalog No. 50-199-4716 Supplier Hach Company Supplier No. 218710
For accuracy checks in calcium hardness determinations.