Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium 2-Hydroxy-4-(methylthio)butyrate, TCI America™
SDP

Catalog No. H065125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H065125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H065125G Supplier TCI America Supplier No. H065125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4857-44-7
Molecular Formula C10H18CaO6S2
Molecular Weight (g/mol) 338.45
MDL Number MFCD00042725
InChI Key ABRVDWASZFDIEH-UHFFFAOYNA-L
Synonym Calcium alpha-Hydroxy-DL-methionine, alpha-Hydroxy-DL-methionine Calcium Salt, 2-Hydroxy-4-(methylthio)butyric Acid Calcium Salt
PubChem CID 107344
IUPAC Name calcium bis(2-hydroxy-4-(methylsulfanyl)butanoate)
SMILES [Ca++].CSCCC(O)C([O-])=O.CSCCC(O)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 338.45
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium 2-Hydroxy-4-(methylthio)butyrate
RTECSNumber ET4731600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.