Learn More
Medchemexpress LLC Calcium 2-hydroxypropanoate pentahydrate | 5743-47-5 | 99.8% | 1 KG
SDP
Catalog No. 5000410556
Encompass_Preferred
Calcium 2-hydroxypropanoate pentahydrate is an activator of hydroxycarboxylic acid receptor 1 (HCAR1) and an epigenetic modulator that induces lactylation of lysine residues. It is a glycolysis end-product that links glycolysis and oxidative phosphorylation, and also functions as an oncometabolite, playing an immune protective role of lactate in anti-tumor immunity.
  • Activator of hydroxycarboxylic acid receptor 1 (HCAR1)
  • Epigenetic modulator that induces lactylation of lysine residues
  • Glycolysis end-product that links glycolysis and oxidative phosphorylation
  • Functions as an oncometabolite
  • Plays an immune protective role of lactate in anti-tumor immunity
Catalog No. 50-004-10556 Supplier Medchemexpress LLC Supplier No. HYB2227D1KG
Calcium 2-hydroxypropanoate pentahydrate is an activator of hydroxycarboxylic acid receptor 1 (HCAR1) and an epigenetic modulator that induces lactylation of lysine residues. It is a glycolysis end-product that links glycolysis and oxidative phosphorylation, and also functions as an oncometabolite, playing an immune protective role of lactate in anti-tumor immunity.
  • Activator of hydroxycarboxylic acid receptor 1 (HCAR1)
  • Epigenetic modulator that induces lactylation of lysine residues
  • Glycolysis end-product that links glycolysis and oxidative phosphorylation
  • Functions as an oncometabolite
  • Plays an immune protective role of lactate in anti-tumor immunity