Learn More
Medchemexpress LLC Calcium 2-hydroxypropanoate pentahydrate | 5743-47-5 | 99.8% | 1 ML
SDP
Catalog No. 5000410641
Encompass_Preferred
Calcium 2-hydroxypropanoate pentahydrate is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. It is also an oncometabolite and has an immune protective role of lactate in anti-tumor immunity.
  • Activates hydroxycarboxylic acid receptor 1 (HCAR1)
  • Induces lysine residues lactylation as an epigenetic modulator
  • Functions as a glycolysis end-product
  • Serves as an oncometabolite
  • Plays an immune protective role of lactate in anti-tumor immunity
Catalog No. 50-004-10641 Supplier Medchemexpress LLC Supplier No. HYB2227D10MM/1ML
Calcium 2-hydroxypropanoate pentahydrate is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. It is also an oncometabolite and has an immune protective role of lactate in anti-tumor immunity.
  • Activates hydroxycarboxylic acid receptor 1 (HCAR1)
  • Induces lysine residues lactylation as an epigenetic modulator
  • Functions as a glycolysis end-product
  • Serves as an oncometabolite
  • Plays an immune protective role of lactate in anti-tumor immunity