Learn More
Selleck Chemical LLC Calcium 2-oxoglutarate S3166-100mg
SDP
Catalog No. 503111600
Encompass_Preferred
Calcium 2-oxoglutarate (Calcium oxoglurate Calcium 2-oxopentanedioate Calcium -ketoglutarate Calcium AKG Calcium 2-ketoglutaric acid Calcium oxoglutaric acid) is the Calcium salt form of 2-oxoglutarate 2-oxoglutarate is naturally found in organisms and is a well-known intermediate in the production of ATP or GTP in the Krebs cycle 2-oxoglutarate is a reversible inhibitor of tyrosinase with IC50 of 15 mM
Catalog No. 50-311-1600 Supplier Selleck Chemical LLC Supplier No. S3166100MG
Calcium 2-oxoglutarate (Calcium oxoglurate Calcium 2-oxopentanedioate Calcium -ketoglutarate Calcium AKG Calcium 2-ketoglutaric acid Calcium oxoglutaric acid) is the Calcium salt form of 2-oxoglutarate 2-oxoglutarate is naturally found in organisms and is a well-known intermediate in the production of ATP or GTP in the Krebs cycle 2-oxoglutarate is a reversible inhibitor of tyrosinase with IC50 of 15 mM