Learn More
Medchemexpress LLC Calcium acetate hydr 1kg
SDP
Catalog No. 5000205401
Encompass_Preferred
Calcium acetate hydrate is a type of calcium salt that can be used as a food additive stabilizer buffer and isolator Calcium acetate hydrate can be combined with phosphate in the diet to reduce phosphate levels in the blood[1]
Catalog No. 50-002-05401 Supplier Medchemexpress LLC Supplier No. HYW0943531KG
Calcium acetate hydrate is a type of calcium salt that can be used as a food additive stabilizer buffer and isolator Calcium acetate hydrate can be combined with phosphate in the diet to reduce phosphate levels in the blood[1]