Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium acetate monohydrat500G
Catalog No. 502897632
Encompass
Calcium acetate monohydrate is calcium salt of acetic acid. Crystals study reveals the structure to be triclinic. Its solubility in aqueous SOLUTIONon over a range of temperature has been studied.
Catalog No. 50-289-7632 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 402850500G
Calcium acetate monohydrate is calcium salt of acetic acid. Crystals study reveals the structure to be triclinic. Its solubility in aqueous SOLUTIONon over a range of temperature has been studied.